Anti-VIP Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-0077R-CY5
10230-562EA 1320 CAD
10230-562
Anti-VIP Rabbit Polyclonal Antibody (Cy5®)
Antibodies
VIP causes vasodilation, lowers arterial blood pressure, stimulates myocardial contractility, increases glycogenolysis and relaxes the smooth muscle of trachea, stomach and gall bladder. PHM and PHV also cause vasodilation. PHM-27 is a potent agonist of the calcitonin receptor CALCR, with similar efficacy as calcitonin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VIP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR