Anti-NKB Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-0070R-CY3
10230-448EA 1300.77 CAD
10230-448
Anti-NKB Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles (By similarity). Is a critical central regulator of gonadal function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NKB
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR