PCR and Gel Clean-up Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN100-0020 GEN100-0050 GEN100-0200
10014-516EA 94.8 CAD
10014-516 10014-518 10014-520
PCR and Gel Clean-up Kit, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Quick, easy kit for purifying DNA from PCR, gels and labeling reactions for a variety of downstream applications.
For the purification of PCR products, extraction of DNA from agarose gels, and removal of nucleotides, primers, enzymes, mineral oil, detergents, and dyes.
PCR & Gel clean-up columns use silica membrane technology in a mini spin column format to deliver easy and efficient DNA purification from PCR products, agarose gels, and labeling reactions. Sample material: Fragment size: 50bp - ~20kbp. Recovery: 70 - 95%. Elution volume: 15 - 50 µl. Preparation time: 10 min for 6 preps. Binding capacity: 25 µg. PCR & Gel clean-up columns have been validated for use with our CGH Labeling Kit, Prod. No ENZ-42671.
Ordering information: Contains Binding buffer (w/ pH indicator), Wash buffer 5x, Elution buffer, PCR & gel clean-up columns, Collection tubes.