Ward's® Laser Pointer
Supplier: AIA / DICK STEIN ADVERTISING
LF1000       
470218-644EA 15.5 CAD
470218-644
Ward's® Laser Pointer
Laser Pointers
Ward's laser pointer is perfect as a presentation aid and for teaching laser optics.
  • Presentation aid
  • Ideal for teaching laser optics
  • Class IIIa red laser
  • Compact, keychain-style
  • Also features an LED flashlight
The compact laser pointer features an LED flashlight and a key chain. Included (not installed) are three long-life LR41 lithium batteries.
Three life-long LR41 lithium batteries are included. To install the batteries, remove them and the non-conductive spacer from the laser pointer, then replace the batteries.
Certifications: FDA and CE approved.