CYTAG® CGH Labeling Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42671-K010 ENZ-42671-K100
10014-576EA 1598.96 CAD
10014-576 10014-578
CYTAG® CGH Labeling Kit, Enzo Life Sciences
Protein Linking and Labeling Systems
Superior labeling efficiency and better dye incorporation results in less failed runs.
Comparative genomic hybridization (aCGH) is a powerful diagnostic tool for detecting DNA copy number gains and losses associated with chromosome abnormalities. aCGH provides an understanding of genetic disorders, cancers and other genomic aberrations. With optimized, proprietary reagents, the Enzo Life Sciences CGH Labeling Kits for oligo arrays produce high quality data using as little as 0.25 µg of genomic DNA, without a need for pre-amplification. For the purification of labeled DNA, use PCR & Gel Clean-up columns, Prod No. ENZ-GEN100.
Ordering information: The 20-reaction kit includes Primers/Reaction Buffer, 1 x 400 µl; Cyanine 3-dUTP Nucleotide Mix, 2 x 50 µl; Cyanine 5-dUTP Nucleotide Mix, 2 x 50 µl; Klenow Exo-DNA Polymerase, 1 x 20 µl; Stop Buffer, 1 x 100 µl; Nuclease-free Water, 1 x 1 ml. The 200-reaction kit includes Primers/Reaction Buffer, 1 x 4 ml; Cyanine 3-dUTP Nucleotide Mix, 1 x 1 ml; Cyanine 5-dUTP Nucleotide Mix, 1 x 1 ml; Klenow Exo-DNA Polymerase, 1 x 0.2 ml; Stop Buffer, 1 x 1 ml; Nuclease-free Water, 1 x 10 ml. Also available in custom high throughput formats. Bulk quantity kit, 2 x 1000 reactions, is available upon special request.