Isometric Goniometers
Supplier: AMERICAN 3B SCIENTIFIC SE
1005903 1009013 1005900
470119-612EA 20 CAD
470119-612 470119-610 470119-600
Isometric Goniometers
Goniometers
These clear plastic goniometers permit the observation of joint's axis of motion and range of motion.
  • Calibrated in 3 scales
  • 360 degree scale
The 360º head has three scales calibrated for use with the ISOM (International Standards of Measurement) system.
Linear scale in inches and centimeters.