Anti-Thrombospondin Mouse Monoclonal Antibody [clone: A6.1]

Supplier: Diagnostic BioSystems
MOB 315-05 MOB 315
CA99990-590EA 4465.38 CAD
CA99990-590 CA95024-832
Anti-Thrombospondin Mouse Monoclonal Antibody [clone: A6.1]
Antibodies
Type:
Antigen: Thrombospondin
Clonality: Monoclonal
Clone: A6.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR