Anti-RPA/p34 Mouse Monoclonal Antibody [clone: 9H8]

Supplier: Diagnostic BioSystems
MOB 329 MOB 329-05
CA99990-616EA 4465.38 CAD
CA99990-616 CA95024-816
Anti-RPA/p34 Mouse Monoclonal Antibody [clone: 9H8]
Antibodies
Type:
Antigen: RPA/p34
Clonality: Monoclonal
Clone: 9H8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR