Anti-MUM1 Mouse Monoclonal Antibody [clone: MUM1p]

Supplier: Diagnostic BioSystems
MOB 420-05 MOB 420
CA95024-692EA 4358.08 CAD
CA95024-692 CA99990-776
Anti-MUM1 Mouse Monoclonal Antibody [clone: MUM1p]
Antibodies
Type:
Antigen: MUM1
Clonality: Monoclonal
Clone: MUM1p
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR