Anti-ACTA1 Mouse Monoclonal Antibody [clone: 5C5]

Supplier: Diagnostic BioSystems
PDM 099
CA99990-978EA 1498.27 CAD
CA99990-978
Anti-ACTA1 Mouse Monoclonal Antibody [clone: 5C5]
Antibodies
Type:
Antigen: ACTA1
Clonality: Monoclonal
Clone: 5C5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR