Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30-F11]

Supplier: ProSci
76-529
10766-450EA 483.75 CAD
10766-450
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30-F11]
Antibodies
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR