Anti-AQP11 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA525704
CAPIPA525704EA 3406.91 CAD
CAPIPA525704
Anti-AQP11 Rabbit Polyclonal Antibody
Antibodies
Aquaporins facilitate the transport of water and small neutral solutes across cell membranes.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: AQP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR