Anti-LAPTM5 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA523585
CAPIPA523585EA 3378.53 CAD
CAPIPA523585
Anti-LAPTM5 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine based on sequence homology.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: LAPTM5 (lysosomal protein transmembrane 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR