3B Scientific® Whiteboard Optics Set
Supplier: American 3B Scientific
1000604 1003321
470162-702EA 1290.39 CAD
470162-702 470162-728
3B Scientific® Whiteboard Optics Set
Educational Materials Physics Educational Materials Optics Learning Activities
Easily Display Optics Principles
  • Kit contains a mixture of reflectors, transparent and shadow objects
  • Use your own light source or the multibeam projector
Set of optical components for use in conjunction with a single-beam lamp or multiple-beam lamp on a white board.
All components are lined with magnetic foil or furnished with a magnet holder and can be easily mounted and aligned on the white board. This apparatus permits a wide range of experiments demonstrating ray optics without the need for a dark room; handwritten notes can be added to provide a clearer explanation.