TNF Alpha (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-099 ADI-901-099
89141-150EA 996.98 CAD
89141-150 89141-330
TNF Alpha (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly sensitive ELISA for measuring TNF-α, the primary mediator of immune regulation.
For the quantitative determination of human TNF-α in culture supernatants, plasma, and serum.
The TNF-α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per 96-well plate. The broad dynamic range makes assaying different pathologies easy.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 21, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.