Cortisol ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
Widely used sensitive cortisol ELISA kit with a large variety of sample types and highly reproducible results for stress, autoimmune disease and cancer research.
For the quantitative determination of Cortisol in culture supernatants, plasma, serum, saliva, and urine from any species. Cited sample type includes feces.
The Cortisol EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit is highly cited in peer-reviewed publications. The ready-to-use liquid color-coded reagents provided are safe, non-radioactive, and reduce error in the lab. The broad dynamic range lets you accurately measure Cortisol levels in a variety of sample matrices.
Ordering information: Contains GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-320 (5x96 wells).