NEBNEXT® rRNA Depletion Kit (Human/Mouse/Rat), New England Biolabs
Supplier: New England Biolabs
E6310S E6310S E6310X E6310L E6310L E6310X
CA102877-570EA 7182 CAD
102877-570 CA102877-570 CA102877-566 CA102877-568 102877-568 102877-566
NEBNEXT® rRNA Depletion Kit (Human/Mouse/Rat), New England Biolabs
Assays Nucleic Acid Assays
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human total RNA preparations
This product is suitable for both intact and degraded RNA (e.g. FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications.