hema-screen™, Immunostics
Supplier: IMMUNOSTICS, INC.
HS-50 HSPP-50 HS-1000 HS-34 HSTR-335CS HS-100
10147-014KT 117.35 CAD
10147-014 10147-020 10147-024 10147-028 10147-030 10147-011
hema-screen™, Immunostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
hema-screen™ is a rapid, convenient, and non-offensive qualitative method for detecting occult blood in the stool
  • Detect hemoglobin as low as 0.6 mg Hb/gm of feces
  • Test time within 30 seconds
Recommended for routine hospital testing and mass screening programs for colorectal cancer detection.
Certifications: CLIA Waived. CE Marked.
Order Now
SPECIFICATIONS
Environmentally Preferable