EDTA (0.5 M), pH 8.0, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-027-101 351-027-721 351-027-721EA
10128-442EA 133 CAD
10128-442 10128-444 10128-446
EDTA (0.5 M), pH 8.0, Quality Biological
Buffers Cloning Buffers
EDTA (Ethylenediaminetetraacetic acid), a metal chelator, is widely used in molecular biology as a chemical reagent to reduce nuclease activity (i.e. metalloenzyme). EDTA solution can be used to study cell biology, molecular biology, bioactive small molecules, biochemicals, solutions and reagents, inorganic salts, and chelators.
"Typically, EDTA is added to Tris base in order to prepare Tris EDTA (TE), which can be used to reconstitute nucleic acids (i.e. DNA).
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.