TE (Tris EDTA) Buffers, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
351-010-131 351-010-721 351-010-721EA 351-011-131 351-011-721 351-011-721EA 351-322-131
10128-412EA 129.39 CAD
10128-412 10128-414 10128-416 10128-418 10128-420 10128-422 10128-586
TE (Tris EDTA) Buffers, Quality Biological
Buffers Cloning Buffers
TE is composed of buffering agent Tris, and chelating agent EDTA. EDTA prevents degradation of DNA and RNA by chelating magnesium (or other divalent metal ions). Together, this Tris-EDTA based solution solubilizes DNA or RNA, while protecting it from degradation.
  • Filtered through a 0.22 µm membrane
  • DNase, RNnase, and Protease tested
The use of TE with slightly different pH values is based on early 80's literature that DNases are supposed to be less active at pH 8.0 and RNases less active at pH 7.5. You can use pH 8.0 for both DNA and RNA.
TE is useful for reconstituting nucleic acids, during DNA precipitation and other DNA applications, storing and diluting DNA and RNA, breaking protein-links in immuno-histochemistry procedures, and unmasking antigens and epitopes in formalin-fixed and paraffin-embedded tissue sections.
Contents: Tris base, disodium EDTA 2H₂O, hydrochloric acid, and Molecular Biology grade water.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.