3B Scientific® Self Breast Exam Trainer With Case
Supplier: American 3B Scientific
1000342 1000343
470119-198EA 4245.6 CAD
470119-198 470119-200
3B Scientific® Self Breast Exam Trainer With Case
Models Human Biology Models
Teach Basic Bse
  • Learn bse in a more natural position
  • Available with a case for easy transport
  • 3 year manufacturers
Demonstrate realistic self-examination with our natural casting of a female upper body with medium sized breasts. It can easily be worn, in order to better train and practice breast self-examination. Displays the skin in finest detail. Very realistic to the touch and dermatologically tested. Breast examination is possible in both upright or lying positions. Benign and malignant tumors in different stages of development hone self-examination skills: 2 benign tumors, 4 malignant tumors, 2 typical anomalies. Includes “Female Breast” chart. Supplied with talcum powder, harness, stand and aluminum carrying case.