3B Scientific® Eye In Bony Orbit
Supplier: American 3B Scientific
1000256
470119-100EA 716.85 CAD
470119-100
3B Scientific® Eye In Bony Orbit
Models Human Biology Models
Removable parts of this anatomical human eye model include: Upper half of the sclera with cornea and eye muscle attachments, both halves of choroid with iris and retina, lens and vitreous humour.
  • 5× life size
  • Includes base
  • Three years manufacturers warranty
  • Includes 3B smart anatomy
This eye model is great for studying the anatomy of the human eye! Eye on base of bony orbit.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.