Serology Kits, Stanbio Laboratory
Supplier: Stanbio Laboratory
1125-050 1125-100 1130-050 1130-100 1155-050 1155-100
10152-608EA 253.08 CAD
10152-608 10152-610 10152-612 10152-614 10152-616 10152-618
Serology Kits, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Our serology kits feature "no dilution" technology which eliminates the need to pre-dilute patient specimens
RaPET® kit includes Latex Reagent, Test Slide, Pipets/Mixers, Dilution Buffer and Controls. Results are Qualitative or Semi-Quantitative.
RPR Quicktest includes RPR Antigen Suspension, Pipets/Stirrers, Dispensing Needle, Dispensing Bottle, 18 mm Circle Test Cards and Three Levels of Controls. Results are Qualitative or Quantitative.
Stanbio Laboratory also offers a charcoal flocculation assay for detecting Syphilis (RPR).
Certifications: All serology tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: The serology test kits comes complete with latex reagent, test slide, pipettes/mixers, dilution buffer and positive and negative controls.