Pregnancy Tests, Stanbio Laboratory
Supplier: Stanbio Laboratory
1220-025 1220-050 1230-025 1230-050 1225-205 1235-305
10152-682EA 309.71 CAD
10152-682 10152-684 10152-686 10152-688 10152-696 10152-698
Pregnancy Tests, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Stanbio Laboratory offers pregnancy tests to meet your clinical and laboratory needs.
Sensivity in Urine 20 mIU/mL
Sensivity in Serum 10 mIU/mL (20 mIU/mL for the True® 20 Plus set)
Certifications: All pregnancy tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: Tests are single step cassette platform formats for urine only or urine/serum pregnancy testing.