3B Scientific® Floral Models
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1017829 1000535 1017828 1017831 1020125 1017832 1000531 1000534 1000532 1000533
470029-550EA 922.1 CAD
470029-550 470311-860 470029-552 470311-864 470103-338 470311-862 470103-346 470029-548 470029-554 470311-858
3B Scientific® Floral Models
Models Botany Models
These floral models illustrate the variety present in basic structure. Choose from economically important plants, like potato, sunflower, apple, cherry and pea to accompany a topic on agricultural Science. Choose sunflower, cherry, or pea for topics of seed type and formation.
If you are discussing the coevolution of plants and insects and the mechanism of pollination, chose the meadow clary and demonstrate the level mechanism inherent in the flower.