Cy™ Amidites
Supplier: Cytiva
28-9172-98 28-9042-49 28-9042-50
CA89128-838EA 720.32 CAD
CA89128-838 CA95040-124L CA95040-126L CA76541-098 CA76541-100 CA76541-102
Cy™ Amidites
Electrophoresis Reagents Western Blotting Reagents
Cy™ fluorescent amidites for incorporation of Cy3, Cy3.5, Cy5, Cy5.5 fluorescent dyes at any position in an oligonucleotide during synthesis.
  • For incorporation of Cy fluorescent dyes at any position in an oligonucleotide during synthesis
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
Cy5 amidite can be used to prepare sequencing primers for use on ALF express.
Delivery information: Larger pack sizes and custom packaging available upon request.