Buffers (10X) For Heat Induced Epitope Recovery, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K043 K044 K038 K 038
CA99990-102EA 617.13 CAD
10064-760 10064-758 CA99990-102 10064-756
Buffers (10X) For Heat Induced Epitope Recovery, Diagnostic BioSystems
Buffers Antibody Purification Buffers
The is a heavy metal free antigen unmasking solution provided in a convenient 10X concentrate
Use of this antigen recovery buffer avoids unnecessary heavy metal exposure to lab personal and handling and disposal issues.