Human Recombinant PLGF 1 (from E. coli)
Supplier: Prosci
40-529
40-151
10072-218EA
840.75
CAD
10072-218
10072-686
Human Recombinant PLGF 1 (from E. coli)
Proteins and Peptides
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, colon and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor and stimulates endothelial cell proliferation and migration. Recombinant human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 131 amino acid polypeptide chains.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.