Human Recombinant NT4 (from E. coli)
Supplier: Prosci
40-498 40-566
10072-230EA 481.9 CAD
10072-230 10073-040
Human Recombinant NT4 (from E. coli)
Proteins and Peptides
NT-4 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-3. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-4 is expressed in the prostate, thymus, placenta and skeletal muscle. NT-4 can signal through the LNGFR and trkB receptors and promotes the survival of peripheral sensory sympathetic neurons. Recombinant human NT-4 is a noncovalently linked homodimer, of two 14.0 kDa polypeptide monomers (260 total amino acid residues). Manufactured using all non-animal reagents.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.