Mouse Recombinant Midkine (from E. coli)

Supplier: Prosci

40-504 40-581
10072-238EA 481.9 CAD
10072-238 10073-064
Mouse Recombinant Midkine (from E. coli)
Proteins and Peptides

Midkine (MK) and its functionally-related protein Pleiotrophin are heparin-binding neurotrophic factors that signal through the same receptor, known as anaplastic lymphoma kinase (ALK). MK plays an important regulatory role in epithelial-mesenchymal interactions during fetal development and in postnatal lung development. MK chemoattracts embryonic neurons, neutrophils and macrophages, and by signaling through the ALK receptor it exerts angiogenic, growth and survival activities during tumorgenesis. Recombinant human Midkine is a 13.4 kDa protein containing 123 amino acid residues including five intra-molecular disulfide bonds. Recombinant murine Midkine is a 13.4 kDa protein containing 121 amino acid residues including five intra-molecular disulfide bonds.


Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Order Now

SPECIFICATIONS

Protein/peptide name Midkine Midkine
Environmentally Preferable
Protein synonyms Amphiregulin-associated protein|ARAP|NEGF2|MK1|Midkine|MK Mek|Retinoic acid-induced differentiation factor|Midkine|Mk|MK
Protein/peptide type Recombinant Recombinant
Species Human Mouse
Source E. coli E. coli
Biological activity Biological activity was determined by its ability to chemoattract human neutrophils using a concentration range of 0.1 - 10.0 ng/mL. Biological activity was determined by its ability to chemoattract human neutrophils using a concentration range of 10-100 ng/mL. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
Purity >98% >98%
Formulation Lyophilized Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR