Mouse Recombinant GMCSF (from E. coli)
Supplier: Prosci
40-593 40-617 40-621 40-475
10070-556EA 840.75 CAD
10070-556 10072-002 10072-010 10072-500
Mouse Recombinant GMCSF (from E. coli)
Proteins and Peptides
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in by endothelial cells, monocytes, fibroblasts and T-lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant rat GM-CSF is a 14.5 kDa globular protein consisting of 128 amino acids residues. Recombinant human GM-CSF is a 14.6 kDa globular protein consisting of 128 amino acids containing two intramolecular disulfide bonds and two potential N-linked glycosylation sites. Recombinant murine GM-CSF (40-617) is a 14.2 kDa globular protein consisting of 124 amino acids residues. Recombinant murine GM-CSF (40-621) is a 14.5 kDa globular protein consisting of 128 amino acids residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.