Mouse Recombinant BMP4 (from E. coli)
Supplier: Prosci
40-203 40-548 40-549 40-615
10072-056EA 481.9 CAD
10072-056 10072-180 10072-182 10073-112
Mouse Recombinant BMP4 (from E. coli)
Proteins and Peptides
Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-β superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body and are involved in diverse physiological processes during both pre- and postnatal life. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva.
Recombinant human BMP-4, expressed in HeLa cells, is a 31-36 kDa homodimeric glycoprotein. Both ProSci’s E.coli derived BMP-4 and PeproTech’s E.coli derived murine BMP-4 are fully active homodimeric protein consisting of two 106 amino acid subunits which correspond to amino acids 303-408 of the full length BMP-4 precursor.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.