Human Recombinant TNF alpha (from E. coli)
Supplier: Prosci
40-446 40-468 40-609
10072-448EA 481.9 CAD
10072-448 10072-486 10073-100
Human Recombinant TNF alpha (from E. coli)
Proteins and Peptides
TNF-alpha is a pleiotrophic pro-inflammatory cytokine secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. It belongs to the TNF family of ligands and signals through two receptors, TNFR1 and TNFR2. TNF-alpha is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-alpha also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. Recombinant murine TNF-alpha is a soluble 156 amino acid protein (17.3 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein. Recombinant human TNF-a is a soluble 157 amino acid protein (17.4 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein. Recombinant rat TNFTNF-a is a soluble 157 amino acid protein (17.3 kDa) which corresponds to C-terminal extracellular domain of the full length transmembrane protein.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.