Rat Recombinant IL-13 (from E. coli)
Supplier: Prosci
40-421 40-469 40-470 40-258
10072-404EA 481.9 CAD
10072-404 10072-488 10072-490 10072-838
Rat Recombinant IL-13 (from E. coli)
Proteins and Peptides
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and also by mast cells and NK cells. Targeted deletion of IL-13 in mice resulted in impaired Th2 cell development and indicated an important role for IL-13 in the expulsion of gastrointestinal parasites. IL-13 exerts anti-inflammatory effects on monocytes and macrophages and it inhibits the expression of inflammatory cytokines such as IL-1β, TNF-α, IL-6 and IL-8. IL-13 has also been shown to enhance B cell proliferation and to induce isotype switching resulting in increased production of IgE. Blocking of IL-13 activity inhibits the pathophysiology of asthma. Human and murine IL-13 is cross-species reactive.
Recombinant murine IL-13 is a 12.3 kDa protein consisting of 111 amino acid residues. Recombinant rat IL-13 (40-469) is an 11.9 kDa protein consisting of 109 amino acid residues. Recombinant rat IL-13 (40-470) is a 12.3 kDa protein consisting of 113 amino acid residues. Recombinant human IL-13 is a 12.5 kDa protein consisting of 114 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.