Mouse Recombinant TRAIL (from E. coli)
Supplier: Prosci
40-459 40-297
10072-472EA 481.9 CAD
10072-472 10072-890
Mouse Recombinant TRAIL (from E. coli)
Proteins and Peptides
TRAIL/Apo2L is a cytotoxic protein, which activates rapid apoptosis in tumor cells, but not in normal cells. TRAIL induced apoptosis is achieved through binding to two death-signaling receptors, DR4 and DR5. These receptors belong to the TNFR superfamily of transmembrane proteins and contain a cytoplasmic "death domain", which activates the cell's apoptotic machinery. Recombinant human soluble TRAIL/Apo2L is a 168 amino acid polypeptide (19.6 kDa), consisting of the TNF homologous portion of the extracellular domain of the full length TRAIL/Apo2L protein.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.