Anti-RETN Chicken Polyclonal Antibody

Supplier: Prosci
54-209
10068-054EA 745.38 CAD
10068-054
Anti-RETN Chicken Polyclonal Antibody
Antibodies
Resistin antibody can be used at 1:2,000 to 1:5,000 for immunoblot or ELISA testings. User needs to optimize the concentrations in assays.

PBS Buffer containing 0.02% sodium azide.

2-8°C for one week and -20°C as an undiluted liquid for up to two years.

Type: Primary
Antigen: Retn
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR