Anti-UNC5CL Goat Polyclonal Antibody

Supplier: Prosci
46-918
10117-284EA 1142.34 CAD
10117-284
Anti-UNC5CL Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 55 kDa band observed in human kidney and ovary lysates (calculated MW of 57.7 kDa according to NP_775832.1). Recommended concentration: 0.3-1 ?g/mL.

UNC5CL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UNC5CL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR