Anti-COL26A1 Rabbit Polyclonal Antibody

Supplier: ProSci
30-837
10110-796EA 1213.98 CAD
10110-796
Anti-COL26A1 Rabbit Polyclonal Antibody
Antibodies
EMID2 contains 1 EMI domain and 2 collagen-like domains. The exact function of ZCCHC3 remains unknown.

EMID2 antibody can be used for detection of EMID2 by ELISA at 1:62500. EMID2 antibody can be used for detection of EMID2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: COL26A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR