Anti-ZNF806 Rabbit Polyclonal Antibody

Supplier: ProSci
30-696
10110-530EA 1213.98 CAD
10110-530
Anti-ZNF806 Rabbit Polyclonal Antibody
Antibodies
The sequence of LOC442049 is derived from an annotated genomic sequence (NW_921562) using gene prediction method: GNOMON. The exact function of LOC442049 remains unknown.

LOC442049 antibody can be used for detection of LOC442049 by ELISA at 1:62500. LOC442049 antibody can be used for detection of LOC442049 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF806
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR