Anti-Troponin I Type 1 Rabbit Polyclonal Antibody

Supplier: Prosci
29-700
10108-744EA 1108.15 CAD
10108-744
Anti-Troponin I Type 1 Rabbit Polyclonal Antibody
Antibodies
Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Each troponin-tropomyosin complex contains 1 molecule from each of the 3 troponins: T, C, and I. The troponin I (TNI) subfamily contains three genes: TNI-skeletal-fast-twitch, TNI-skeletal-slow-twitch, and TNI-cardiac. TNNI1 prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.

TNNI1 antibody can be used for detection of TNNI1 by ELISA at 1:62500. TNNI1 antibody can be used for detection of TNNI1 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNNI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat. Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR