Anti-ZNF286A Rabbit Polyclonal Antibody

Supplier: ProSci
29-060
10107-528EA 1108.15 CAD
10107-528
Anti-ZNF286A Rabbit Polyclonal Antibody
Antibodies
ZNF286 may be involved in transcriptional regulation.

ZNF286 antibody can be used for detection of ZNF286 by ELISA at 1:312500. ZNF286 antibody can be used for detection of ZNF286 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF286A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR