Anti-HOXC10 Rabbit Polyclonal Antibody

Supplier: ProSci
29-036
10107-480EA 1213.98 CAD
10107-480
Anti-HOXC10 Rabbit Polyclonal Antibody
Antibodies
HOXC10 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation.

HOXC10 antibody can be used for detection of HOXC10 by ELISA at 1:12500. HOXC10 antibody can be used for detection of HOXC10 by western blot at 0.0625 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HOXC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR