Anti-NS1BP Rabbit Polyclonal Antibody

Supplier: Prosci
27-789
10105-488EA 1097.39 CAD
10105-488
Anti-NS1BP Rabbit Polyclonal Antibody
Antibodies
IVNS1ABP is a novel human protein that interacts with the influenza A virus. Nonstructural NS1 protein is relocalized in the nuclei of infected cells.

IVNS1ABP antibody can be used for detection of IVNS1ABP by ELISA at 1:312500. IVNS1ABP antibody can be used for detection of IVNS1ABP by western blot at 2.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IVNS1ABP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR