PermaGreen Plus/AP, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K059-110 K059 K059
CA10064-832EA 307.83 CAD
10064-766 10064-832 CA10064-832
PermaGreen Plus/AP, Diagnostic BioSystems
Stains Electrophoresis Stains
PermaGreen Plus/AP is a substrate-chromogen system designed for either IHC or ISH when utilizing alkaline phosphatase
It shows increased staining intensity, and produces a strong green color that is insoluble in alcohol and xylene substitutes therefore sections can be dehydrated in alcohol, cleared in a xylene substitute, and permanently mounted.