IMAP Kinase, Phosphatase, and Phosphodiesterase Assays, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
R8155 R8161 R8175 R8176 R8127 R8160 R8155 R8161 R8175 R8176 R8073 R8127 R8160
10048-838EA 1267.51 CAD
10048-838 10048-840 10048-842 10048-844 10048-846 10048-848 10048-850 CA10048-838 CA10048-840 CA10048-842 CA10048-844 CA10048-846 CA10048-848 CA10048-850
IMAP Kinase, Phosphatase, and Phosphodiesterase Assays, Molecular Devices
Assays Enzyme Assays
IMAP® technology provides a homogeneous assay applicable to a wide variety of kinases without regard for substrate peptide sequence.
  • Complete assay system
  • Robust fluorescence signal
  • Homogeneous
  • Available in both FP and TR-FRET detection modes
IMAP provides a complete assay system for screening kinases, phosphatases, and phosphodiesterases
The assay is a simple "mix-and-read" procedure that allows accurate determination of kinase, phosphatase, and phosphodiesterase (PDE) activity.