Miriad Rapid TP/HIV Test LAB+, MedMira
Supplier: MedMira
Miriad™
815311005961 815311005961
CA95062-813PK 1191.9 CAD
10847-594 CA95062-813
Miriad Rapid TP/HIV Test LAB+, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid TP/HIV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of syphilis (Treponema pallidum bacteria) and HIV-1/2
Miriad TP/HIV features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps users can obtain instant results on a single test cartridge.
Ordering information: Each box of Miriad TP/HIV includes 50 text kits.