Miriad Rapid HBc/HIV/HCV Antibody Test, LAB+ Format, MedMira
Supplier: MEDMIRA CA
815311005978
CA95062-809PK 1191.9 CAD
CA95062-809
Miriad Rapid HBc/HIV/HCV Antibody Test, LAB+ Format, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid HBc/HIV/HCV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis B core, HIV-1/2, and Hepatitis C.
Miriad HBc/HIV/HCV features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for up to 18 months at 2 to 30 °C (35.6 to 86 °F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
Ordering information: HBc/HIV/HCV Antibody Test Controls are available as an accessory to the test (10769-911).
Delivery information: Each box of Miriad HBc/HIV/HCV LAB+ Format includes 50 test kits, 50 sample prep tubes, 50 transfer pipettes and 2×30 ml bottles of universal buffer.