Miriad RVF Toolkit is a set of components and protocols based on MedMira Rapid Vertical Flow Technology enabling researchers and academics with the building blocks to create and optimize assays comparable to the complex laboratory-based testing systems
The Toolkit features instant assay results, simultaneous detection of multiple biomarkers on a single device, and room temperature storage. Sample types and volumes are flexible and no specialized equipment is required. Users of the Miriad Toolkit also have access to MedMira consultants and licensing agreements. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
Researchers can quickly detect an analyte of interest – 1) spot the antigen on the membrane, 2) let it dry, and 3) run the test. Users can also use Miriad to monitor bioreactors or simply improve lab processes. Miriad is a simple migration route for researchers to move existing lab tests to rapid tests to be used in-house or commercialized.
Certifications: UL Listed
Ordering information: Each Miriad RVF Toolkit includes: 50 test cartridges, 50 InstantGold Caps, 2 bottles (30mL) of Universal Buffer, 1 bag of disposable pipettes, 1 bottle (5 mL) of Concentrated Gold, and 1 Product Guide.