Oil Red O Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
KT025 KT 025
CA95027-208EA 1920.47 CAD
95027-208 CA95027-208
Oil Red O Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains fat cells and neutral fat in frozen tissue sections
The fat cells and neutral fat stains red, while the nuclei stain blue.