Homogenizer Plates, E-Z 96®, and Spin Columns, Omega Bio-Tek
Supplier: Omega Bio-Tek
HCR003 HCR9601-02 HCR000
CA101414-438CS 398 CAD
CA101414-438 CA101414-440 CA101414-442 CA76539-246
Homogenizer Plates, E-Z 96®, and Spin Columns, Omega Bio-Tek
Microplates Filter Plates
For simple and rapid homogenisation of cell and tissue lysates.
The E-Z 96® homogeniser plates and E.Z.N.A.® spin columns provide a simple and reliable method to reduce sample viscosity often caused by high molecular weight cellular components and cell debris during DNA/RNA isolation. Traditional methods use syringes and needles, which can be tedious, inconvenient, and hazardous to handle. The E-Z 96® homogeniser plates and E.Z.N.A.® spin columns consist of a unique biopolymer-shredding membrane in spin column or 96-well format. In general, similar yields and RNA quality are obtained as with rotor-stator homogenisation.